missing translation for 'onlineSavingsMsg'
Learn More

1-(2,6-Dichlorophenyl)oxindole 98.0+%, TCI America™

$88.56 - $271.44

Chemical Identifiers

CAS 15362-40-0
Molecular Formula C14H9Cl2NO
Molecular Weight (g/mol) 278.13
MDL Number MFCD00451393
InChI Key JCICIFOYVSPMHG-UHFFFAOYSA-N
Synonym 1-(2,6-Dichlorophenyl)-2-oxoindoline, 1-(2,6-Dichlorophenyl)-2-indolinone, Diclofenac Amide
PubChem CID 27211
IUPAC Name 1-(2,6-dichlorophenyl)-2,3-dihydro-1H-indol-2-one
SMILES ClC1=CC=CC(Cl)=C1N1C(=O)CC2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D42605G
View Documents
TCI America
D42605G
5 g
Each for $88.56
Only null left
 
D426025G
View Documents
TCI America
D426025G
25 g
Each for $271.44
Only null left
 
Description

Description

Specifications

Chemical Identifiers

15362-40-0
278.13
JCICIFOYVSPMHG-UHFFFAOYSA-N
27211
ClC1=CC=CC(Cl)=C1N1C(=O)CC2=CC=CC=C12
C14H9Cl2NO
MFCD00451393
1-(2,6-Dichlorophenyl)-2-oxoindoline, 1-(2,6-Dichlorophenyl)-2-indolinone, Diclofenac Amide
1-(2,6-dichlorophenyl)-2,3-dihydro-1H-indol-2-one

Specifications

15362-40-0
C14H9Cl2NO
5 g
JCICIFOYVSPMHG-UHFFFAOYSA-N
1-(2,6-dichlorophenyl)-2,3-dihydro-1H-indol-2-one
27211
≥98.0% (GC)
1-(2,6-Dichlorophenyl)oxindole
127°C
MFCD00451393
1-(2,6-Dichlorophenyl)-2-oxoindoline, 1-(2,6-Dichlorophenyl)-2-indolinone, Diclofenac Amide
ClC1=CC=CC(Cl)=C1N1C(=O)CC2=CC=CC=C12
278.13
278.13
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : NM3259200

TSCA : No

SDS
Documents

Documents