accessibility menu, dialog, popup

UserName

CAS RN 15362-40-0

ClClNO

CAS RN 15362-40-0

IUPAC Name: 1-(2,6-dichlorophenyl)-2,3-dihydro-1H-indol-2-one
Synonyms: 1-(2,6-dichlorophenyl)-3H-indol-2-one
Molecular Weight (g/mol): 278.13
Molecular Formula: C14H9Cl2NO
InChi Key: JCICIFOYVSPMHG-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Cl)=C1N1C(=O)CC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results