missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2,6-Dichlorophenyl)oxindole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 127°C |
| Quantity | 5 g |
| Formula Weight | 278.13 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-(2,6-Dichlorophenyl)oxindole |
Safety and Handling
| RTECSNumber | NM3259200 |
| TSCA | No |