missing translation for 'onlineSavingsMsg'
Learn More

1,2,5-Trimethylpyrrole 98.0+%, TCI America™

Catalog No. T19395ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19395ML 5 mL
T193925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19395ML Supplier TCI America Supplier No. T19395ML
Only null left

Chemical Identifiers

CAS 930-87-0
Molecular Formula C7H11N
Molecular Weight (g/mol) 109.172
MDL Number MFCD00003090
InChI Key YRABRACUKBOTKB-UHFFFAOYSA-N
Synonym 1,2,5-trimethyl-1h-pyrrole, 1h-pyrrole, 1,2,5-trimethyl, pyrrole, 1,2,5-trimethyl, acmc-209rjx, 1,2,5-trimehylpyrrole, 1,2,5-trimethylpyrrole, 1,2,5-trimethyl-1h-pyrrole #, 1,2,5-trimethyl-3h-pyrrole-1-ium-3-ide
PubChem CID 70260
IUPAC Name 1,2,5-trimethylpyrrole
SMILES CC1=CC=C(N1C)C

Specifications

Color Yellow
Quantity 5 mL
UN Number 1993
Formula Weight 109.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,5-Trimethylpyrrole