missing translation for 'onlineSavingsMsg'
Learn More

1,2,5-Pentanetriol 97.0+%, TCI America™

Catalog No. P13505G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13505G 5 g
P13501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13505G Supplier TCI America Supplier No. P13505G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14697-46-2
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.15
MDL Number MFCD01709258
InChI Key WEAYWASEBDOLRG-UHFFFAOYNA-N
PubChem CID 85763
IUPAC Name pentane-1,2,5-triol
SMILES OCCCC(O)CO

Specifications

Color Yellow
Boiling Point 195°C
Quantity 5 g
Formula Weight 120.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,5-Pentanetriol
RTECSNumber SA3327000
TSCA No