CAS RN 14697-46-2
CAS RN 14697-46-2
IUPAC Name:
pentane-1,2,5-triol
Synonyms:
pentane-1,2,5-triol
Molecular Weight (g/mol):
120.15
Molecular Formula:
C5H12O3
InChi Key:
WEAYWASEBDOLRG-UHFFFAOYNA-N
SMILES:
OCCCC(O)CO
1
–
1
of
1
results
1
1,2,5-Pentanetriol 97.0+%, TCI America™
CAS: 14697-46-2 Molecular Formula: C5H12O3 Molecular Weight (g/mol): 120.15 MDL Number: MFCD01709258 InChI Key: WEAYWASEBDOLRG-UHFFFAOYNA-N PubChem CID: 85763 IUPAC Name: pentane-1,2,5-triol SMILES: OCCCC(O)CO