missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trivinylcyclohexane (mixture of isomers) 97.0+%, TCI America™

Catalog No. T089925ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T089925ML 25 mL
T0899500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T089925ML Supplier TCI America Supplier No. T089925ML

Chemical Identifiers

CAS 2855-27-8
Molecular Formula C12H18
Molecular Weight (g/mol) 162.28
MDL Number MFCD00001515
InChI Key KTRQRAQRHBLCSQ-UHFFFAOYNA-N
PubChem CID 96529
IUPAC Name 1,2,4-triethenylcyclohexane
SMILES C=CC1CCC(C=C)C(C1)C=C

Specifications

Color Yellow
Boiling Point 88°C
Quantity 25 mL
Formula Weight 162.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,4-Trivinylcyclohexane (mixture of isomers)