missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trimethylcyclohexane (mixture of stereoisomers) 96.0+%, TCI America™

Catalog No. p-7136446
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
T082525ML 25 mL
T0825500ML 500 mL
2 options

Catalog No. T082525ML

Supplier: TCI America T082525ML

Only null left

Chemical Identifiers

CAS 2234-75-5
Molecular Formula C9H18
Molecular Weight (g/mol) 126.24
MDL Number MFCD00019385
InChI Key VCJPCEVERINRSG-UHFFFAOYNA-N
PubChem CID 91517
IUPAC Name 1,2,4-trimethylcyclohexane
SMILES CC1CCC(C)C(C)C1

Specifications

Color Yellow
Boiling Point 145°C
Quantity 25 mL
UN Number 3295
Formula Weight 126.24
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,4-Trimethylcyclohexane (mixture of stereoisomers)