accessibility menu, dialog, popup

UserName

CAS RN 2234-75-5

CH3CH3H3C

CAS RN 2234-75-5

IUPAC Name: 1,2,4-trimethylcyclohexane
Synonyms: 1,2,4-trimethylcyclohexane
Molecular Weight (g/mol): 126.24
Molecular Formula: C9H18
InChi Key: VCJPCEVERINRSG-UHFFFAOYNA-N
SMILES: CC1CCC(C)C(C)C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results