missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trimethylcyclohexane (mixture of stereoisomers) 96.0+%, TCI America™

Catalog No. T082525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T082525ML 25 mL
T0825500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T082525ML Supplier TCI America Supplier No. T082525ML
Only null left

Chemical Identifiers

CAS 2234-75-5
Molecular Formula C9H18
Molecular Weight (g/mol) 126.24
MDL Number MFCD00019385
InChI Key VCJPCEVERINRSG-UHFFFAOYNA-N
PubChem CID 91517
IUPAC Name 1,2,4-trimethylcyclohexane
SMILES CC1CCC(C)C(C)C1

Specifications

Color Yellow
Boiling Point 145°C
Quantity 25 mL
UN Number 3295
Formula Weight 126.24
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,4-Trimethylcyclohexane (mixture of stereoisomers)