missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trimethylbenzene 98.0+%, TCI America™

Catalog No. p-7136300
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T046925ML 25 mL
T0469500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T046925ML Supplier TCI America Supplier No. T046925ML
Only null left

Chemical Identifiers

CAS 95-63-6
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00008527
InChI Key GWHJZXXIDMPWGX-UHFFFAOYSA-N
Synonym pseudocumene, pseudocumol, psi-cumene, as-trimethylbenzene, 1,3,4-trimethylbenzene, benzene, 1,2,4-trimethyl, uns-trimethylbenzene, 1,2,5-trimethylbenzene, asymmetrical trimethylbenzene, .psi.-cumene
PubChem CID 7247
ChEBI CHEBI:34039
IUPAC Name 1,2,4-trimethylbenzene
SMILES CC1=CC(=C(C=C1)C)C

Specifications

Color Colorless
Boiling Point 169°C
Quantity 25 mL
UN Number 3295
Formula Weight 120.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,4-Trimethylbenzene
EINECSNumber (3)-0007&(3)-3427
RTECSNumber DC3325000
TSCA Yes