accessibility menu, dialog, popup

UserName

CH3CH3H3C

CAS RN 95-63-6

IUPAC Name: 1,2,4-trimethylbenzene
Synonyms: 1,2,4-trimethylbenzene uns-trimethylbenzene pseudocumol asymmetrical trimethylbenzene as-trimethylbenzene trimethylbenzene
Molecular Weight (g/mol): 120.2
Molecular Formula: C9H12
InChi Key: GWHJZXXIDMPWGX-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(C)=C1
Boiling Point 
  • (1)
  • (3)
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (3)
  • (7)
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (7)
  • (1)
  • (3)
  • (2)
  • (4)
  • (3)
  • (7)
  • (2)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (9)