missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trimethoxybenzene 97.0+%, TCI America™

Catalog No. T113025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T113025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T113025G Supplier TCI America Supplier No. T113025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135-77-3
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00008360
InChI Key AGIQIOSHSMJYJP-UHFFFAOYSA-N
Synonym benzene, 1,2,4-trimethoxy, unii-3ku3wd07ss, epa pesticide chemical code 040515, 3ku3wd07ss, hydroxyhydroquinone trimethyl ether, 2,4,5-trimethoxybenzene, acmc-1br3s, 1,3,4-trimethoxy benzene, dsstox_cid_15163, dsstox_rid_79243
PubChem CID 67284
IUPAC Name 1,2,4-trimethoxybenzene
SMILES COC1=CC(=C(C=C1)OC)OC

Specifications

Boiling Point 247°C
Quantity 25 g
Formula Weight 168.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,4-Trimethoxybenzene