missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trimethoxybenzene, 98%

Catalog No. AAA1048318
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1048318 50 g
AAA1048309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1048318 Supplier Thermo Scientific Chemicals Supplier No. A1048318
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 135-77-3 | C9H12O3 | 168.192 g/mol

1,2,4-Trimethoxybenzene is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4-Trimethoxybenzene is used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Store away from incompatible materials such as oxidizing agents, acids and bases. Use only in a chemical fumehood.

Chemical Identifiers

CAS 135-77-3
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00008360
InChI Key AGIQIOSHSMJYJP-UHFFFAOYSA-N
Synonym benzene, 1,2,4-trimethoxy, unii-3ku3wd07ss, epa pesticide chemical code 040515, 3ku3wd07ss, hydroxyhydroquinone trimethyl ether, 2,4,5-trimethoxybenzene, acmc-1br3s, 1,3,4-trimethoxy benzene, dsstox_cid_15163, dsstox_rid_79243
PubChem CID 67284
IUPAC Name 1,2,4-trimethoxybenzene
SMILES COC1=CC(=C(C=C1)OC)OC

Specifications

Density 1.128
Boiling Point 247°C
Flash Point >110°C (230°F)
Refractive Index 1.533
Quantity 50 g
Beilstein 2047579
Merck Index 14,1073
Solubility Information Not miscible in water.
Formula Weight 168.19
Percent Purity 98%
Chemical Name or Material 1,2,4-Trimethoxybenzene
Show More Show Less
EINECSNumber 205-219-7
RTECSNumber DC2770000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only