missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trifluorobenzene 98.0+%, TCI America™

Catalog No. p-7136564
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10335G 5 g
T103325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10335G Supplier TCI America Supplier No. T10335G
Only null left

Chemical Identifiers

CAS 367-23-7
Molecular Formula C6H3F3
Molecular Weight (g/mol) 132.09
MDL Number MFCD00000305
InChI Key PEBWOGPSYUIOBP-UHFFFAOYSA-N
Synonym benzene, 1,2,4-trifluoro, unii-qww4hnq0l1, 1,3,4-trifluorobenzene, qww4hnq0l1, pubchem1068, 1,4-trifluorobenzene, benzene,2,4-trifluoro, 1,2,4-trifluoro-benzene, 1,2,4-trifluorobenzene, acmc-209io5
PubChem CID 67773
IUPAC Name 1,2,4-trifluorobenzene
SMILES FC1=CC=C(F)C(F)=C1

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
UN Number 1993
Formula Weight 132.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,4-Trifluorobenzene