missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,4-Trichlorobenzene, CHROMASOLV™, ≥99%, Honeywell Riedel-de Haën™
CHROMASOLV™,≥99%
Supplier: Honeywell-Chromasolv 2564122L
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
| 1,2,4-Trichlorobenzene | |
| CHROMASOLV™, ≥99% | |
| ≤100.0000% | |
| 213.5°C | |
| 105°C | |
| C6H3Cl3 | |
| MFCD00000547 | |
| 956819 | |
| PBKONEOXTCPAFI-UHFFFAOYSA-N | |
| 1,2,4-trichlorobenzene | |
| 13 | |
| 181.46 | |
| HPLC |
| 16°C (literature) | |
| 120-82-1 | |
| 1.454Â g/mLÂ at 25Â °C(lit.) | |
| 2 L | |
| Glass Bottle | |
| C6H3Cl3 | |
| UN2321 | |
| benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene | |
| C1=CC(=C(C=C1Cl)Cl)Cl | |
| 181.44 | |
| CHEBI:28222 | |
| >99% |
Chemical Identifiers
| 120-82-1 | |
| 181.44 | |
| PBKONEOXTCPAFI-UHFFFAOYSA-N | |
| 13 | |
| 1,2,4-trichlorobenzene |
| C6H3Cl3 | |
| MFCD00000547 | |
| benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene | |
| CHEBI:28222 | |
| C1=CC(=C(C=C1Cl)Cl)Cl |
Safety and Handling
ShelfLife : 1620 days from date of manufacture
DOTInformation : Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Trichlorobenzenes, liquid
EINECSNumber : 204-428-0
Recommended Storage : Room Temp