missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trichlorobenzene, CHROMASOLV™, ≥99%, Honeywell Riedel-de Haën™

Catalog No. p-7111777
Change view
Click to view available options
Quantity:
1 L
2 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045708 1 L Glass Bottle
6000070 2 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60045708 Supplier Honeywell Chemicals Supplier No. 2564121L
Only null left

CHROMASOLV™,≥99%

  • High purity chromatography solvents
  • Suitability tested for specified use from HPLC up to UHPLC-MS applications.

Chemical Identifiers

CAS 120-82-1
Molecular Formula C6H3Cl3
Molecular Weight (g/mol) 181.44
MDL Number MFCD00000547
InChI Key PBKONEOXTCPAFI-UHFFFAOYSA-N
Synonym benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene
PubChem CID 13
ChEBI CHEBI:28222
IUPAC Name 1,2,4-trichlorobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)Cl

Specifications

Melting Point 16°C (literature)
CAS Max % ≤100.0000%
Density 1.454 g/mL at 25 °C(lit.)
Quantity 1 L
Flash Point 105°C
Packaging Glass Bottle
Linear Formula C6H3Cl3
UN Number UN2321
Beilstein 956819
Boiling Point 213.5°C
Formula Weight 181.46
Percent Purity >99%
Grade HPLC
Chemical Name or Material 1,2,4-Trichlorobenzene, CHROMASOLV™, ≥99%
Show More Show Less
ShelfLife 1620 days from date of manufacture
DOTInformation Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Trichlorobenzenes, liquid
EINECSNumber 204-428-0
Recommended Storage Room Temp