missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Tribromobenzene 95.0+%, TCI America™

Catalog No. T03465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T03465G 5 g
T034625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T03465G Supplier TCI America Supplier No. T03465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 615-54-3
Molecular Formula C6H3Br3
Molecular Weight (g/mol) 314.802
MDL Number MFCD00000062
InChI Key FWAJPSIPOULHHH-UHFFFAOYSA-N
PubChem CID 12002
IUPAC Name 1,2,4-tribromobenzene
SMILES C1=CC(=C(C=C1Br)Br)Br

Specifications

Melting Point 42°C
Color White-Yellow
Quantity 5 g
Synonym benzene, 1,2,4-tribromo, unii-eoe1kfc260, benzene,1,2,4-tribromo, eoe1kfc260, dsstox_cid_4346, dsstox_rid_77377, dsstox_gsid_24346, 1,2,4-tribomobenzene, benzene,2,4-tribromo, acmc-1bdp2
Formula Weight 314.80
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,2,4-Tribromobenzene
RTECSNumber DC1956500
TSCA Yes