missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Tribromobenzene, 95%

CAS: 615-54-3 | C6H3Br3 | 314.802 g/mol

$79.79 - $233.01

Chemical Identifiers

CAS 615-54-3
Molecular Formula C6H3Br3
Molecular Weight (g/mol) 314.802
MDL Number MFCD00000062
InChI Key FWAJPSIPOULHHH-UHFFFAOYSA-N
PubChem CID 12002
IUPAC Name 1,2,4-tribromobenzene
SMILES C1=CC(=C(C=C1Br)Br)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2377806
View Documents
Thermo Scientific Chemicals
B2377806
5 g
Each for $79.79
Only null left
 
AAB2377814
View Documents
Thermo Scientific Chemicals
B2377814
25 g
Each for $233.01
Only null left
 
Description

Description

1,2,4-tribromobenzene is used in the synthesis of hyper branched poly(p-phenylene ethynylenes). It is used as cross-linking reagent during Pd-catalyzed cross-coupling of 2,5-diiodo-4-[(2-ethylhexyl)oxy]methoxybenzene and 1,4-diethynyl-2,5-bis-(octyloxy)benzene. It is a compound commonly used in organic synthesis reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4-tribromobenzene is used in the synthesis of hyper branched poly(p-phenylene ethynylenes). It is used as cross-linking reagent during Pd-catalyzed cross-coupling of 2,5-diiodo-4-[(2-ethylhexyl)oxy]methoxybenzene and 1,4-diethynyl-2,5-bis-(octyloxy)benzene. It is a compound commonly used in organic synthesis reactions.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

615-54-3
314.802
FWAJPSIPOULHHH-UHFFFAOYSA-N
1,2,4-tribromobenzene
C6H3Br3
MFCD00000062
12002
C1=CC(=C(C=C1Br)Br)Br

Specifications

615-54-3
274°C to 276°C
C6H3Br3
5 g
benzene, 1,2,4-tribromo, unii-eoe1kfc260, benzene,1,2,4-tribromo, eoe1kfc260, dsstox_cid_4346, dsstox_rid_77377, dsstox_gsid_24346, 1,2,4-tribomobenzene, benzene,2,4-tribromo, acmc-1bdp2
FWAJPSIPOULHHH-UHFFFAOYSA-N
1,2,4-tribromobenzene
12002
95%
41°C to 43°C
>110°C (230°F)
MFCD00000062
1934823
Insoluble in water.
C1=CC(=C(C=C1Br)Br)Br
314.802
314.8
1,2,4-Tribromobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335-H500

EINECSNumber : 210-433-9

RTECSNumber : DC1956500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only