missing translation for 'onlineSavingsMsg'
Learn More

[1,2,4]Triazolo[4,3-a]pyridine-3-thiol, 97%, Thermo Scientific™

Catalog No. MO07000DA
Change view
Click to view available options
Quantity:
1g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO07000DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07000DA Supplier Thermo Scientific Chemicals Supplier No. MO07000DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6952-68-7
Molecular Formula C6H5N3S
Molecular Weight (g/mol) 151.19
MDL Number MFCD03783345
InChI Key ZQMDNIPQCWNIMG-UHFFFAOYSA-N
Synonym 1,2,4 triazolo 4,3-a pyridine-3-thiol, 1,2,4-triazolo 4,3-a pyridine-3 2h-thione, s-triazolo 4,3-a pyridine-3 2h-thione, unii-t0rly3osj5, t0rly3osj5, 2h-1,2,4 triazolo 4,3-a pyridine-3-thione, 1,2,4 triazolo 4,3-a pyridine-3 2h-thione, 3-mercaptotriazolo 4,3-a pyridine, 1,2,4-triazolo 4,3-a pyridine-3-thiol
PubChem CID 676422
IUPAC Name 2H,3H-[1,2,4]triazolo[4,3-a]pyridine-3-thione
SMILES S=C1NN=C2C=CC=CN12

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 151.19
Percent Purity 97%
Chemical Name or Material [1,2,4]Triazolo[4,3-a]pyridine-3-thiol
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.