missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Triazolo[4,3-a]pyridine-3-thiol, 96%

Catalog No. AAH3269403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3269403 1 g
AAH3269406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3269403 Supplier Thermo Scientific Chemicals Supplier No. H3269403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6952-68-7 | C6H5N3S | 151.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6952-68-7
Molecular Formula C6H5N3S
Molecular Weight (g/mol) 151.19
MDL Number MFCD03783345
InChI Key ZQMDNIPQCWNIMG-UHFFFAOYSA-N
Synonym 1,2,4 triazolo 4,3-a pyridine-3-thiol, 1,2,4-triazolo 4,3-a pyridine-3 2h-thione, s-triazolo 4,3-a pyridine-3 2h-thione, unii-t0rly3osj5, t0rly3osj5, 2h-1,2,4 triazolo 4,3-a pyridine-3-thione, 1,2,4 triazolo 4,3-a pyridine-3 2h-thione, 3-mercaptotriazolo 4,3-a pyridine, 1,2,4-triazolo 4,3-a pyridine-3-thiol
PubChem CID 676422
IUPAC Name 2H,3H-[1,2,4]triazolo[4,3-a]pyridine-3-thione
SMILES S=C1NN=C2C=CC=CN12

Specifications

Melting Point 209°C to 210°C
Quantity 1 g
Formula Weight 151.19
Percent Purity 96%
Chemical Name or Material 1,2,4-Triazolo[4,3-a]pyridine-3-thiol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only