missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Triazole, sodium salt, 90%, technical

Catalog No. p-202121
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC188990100 10 g Glass bottle
AC188990500 50 g Glass bottle
AC188992500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC188990100 Supplier Thermo Scientific Chemicals Supplier No. 188990100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 41253-21-8 | C2H2N3Na | 91.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 41253-21-8
Molecular Formula C2H2N3Na
Molecular Weight (g/mol) 91.05
MDL Number MFCD00044437
InChI Key NVMNEWNGLGACBB-UHFFFAOYSA-N
Synonym sodium 1,2,4-triazol-1-ide, 1,2,4-triazolylsodium, 1,2,4-triazole sodium derivative, 1,2,4-triazole sodium salt, sodium 1,2,4-triazolide, 1h-1,2,4-triazole, sodium salt, sodium 1h-1,2,4-triazolate, 1,2,4-triazole sodium, sodium 1h-1,2,4-triazolide, 1,2,4-triazole, sodium salt
PubChem CID 4649906
IUPAC Name sodium;2,4-diaza-1-azanidacyclopenta-2,4-diene
SMILES C1=NC=N[N-]1.[Na+]

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 295°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Quantity 10 g
Beilstein 26, II, 7
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in most organic solvents
Formula Weight 91.05
Percent Purity 90%
Grade Technical
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4-Triazole, sodium derivative, tech., 90%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 255-280-9
TSCA TSCA

RUO – Research Use Only