missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Triacetoxybenzene 95.0+%, TCI America™

Catalog No. p-7136529
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T097625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T097625G Supplier TCI America Supplier No. T097625G
Only null left

Chemical Identifiers

CAS 613-03-6
Molecular Formula C12H12O6
Molecular Weight (g/mol) 252.222
MDL Number MFCD00008700
InChI Key AESFGSJWSUZRGW-UHFFFAOYSA-N
Synonym 1,2,4-triacetoxybenzene, benzene-1,2,4-triyl triacetate, hydroxyhydroquinone triacetate, 1,2,4-benzenetriol, triacetate, pyrogallol a, 2-hydroxyhydroquinone triacetate, 1,2,4-benzenetriacetate, unii-903t7h7cp1, triacetate d'hydroxyhydroquinone, 2,4-bis acetyloxy phenyl acetate
PubChem CID 69169
IUPAC Name (3,4-diacetyloxyphenyl) acetate
SMILES CC(=O)OC1=CC(=C(C=C1)OC(=O)C)OC(=O)C

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 25 g
Formula Weight 252.22
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4-Triacetoxybenzene
RTECSNumber DC4800000
TSCA Yes