missing translation for 'onlineSavingsMsg'
Learn More

1-(2,4-Dimethylphenyl)ethanol, 95%

Catalog No. AAH3214109
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3214109 10 g
AAH3214103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3214109 Supplier Thermo Scientific Chemicals Supplier No. H3214109
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5379-19-1 | C10H14O | 150.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5379-19-1
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00060890
InChI Key DNHQUGRUHBFDFT-UHFFFAOYSA-N
Synonym 1-2,4-dimethylphenyl ethanol, 1-2,4-dimethylphenyl ethan-1-ol, 1-2,4-dimethylbenzene-1-ethanol
PubChem CID 21475
IUPAC Name 1-(2,4-dimethylphenyl)ethanol
SMILES CC1=CC(=C(C=C1)C(C)O)C

Specifications

Quantity 10 g
Formula Weight 150.22
Percent Purity 95%
Chemical Name or Material 1-(2,4-Dimethylphenyl)ethanol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only