missing translation for 'onlineSavingsMsg'
Learn More

1-(2,4-Difluorophenyl)piperazine, 99%, Thermo Scientific™

$73.78 - $744.96

Chemical Identifiers

CAS 115761-79-0
Molecular Formula C10H12F2N2
Molecular Weight (g/mol) 198.217
MDL Number MFCD00082588
InChI Key CMCSPBOWEYUGHB-UHFFFAOYSA-N
PubChem CID 2734637
IUPAC Name 1-(2,4-difluorophenyl)piperazine
SMILES C1CN(CCN1)C2=C(C=C(C=C2)F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1939303
View Documents
Thermo Scientific Chemicals
L1939303
1 g
Each for $73.78
Only null left
 
AAL1939306
View Documents
Thermo Scientific Chemicals
L1939306
5 g
Each for $206.53
Only null left
 
AAL1939314
View Documents
Thermo Scientific Chemicals
L1939314
25 g
Each for $744.96
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

115761-79-0
198.217
CMCSPBOWEYUGHB-UHFFFAOYSA-N
1-(2,4-difluorophenyl)piperazine
C10H12F2N2
MFCD00082588
2734637
C1CN(CCN1)C2=C(C=C(C=C2)F)F

Specifications

115761-79-0
108°C to 111°C (0.2 mmHg)
MFCD00082588
UN2923
Air Sensitive
CMCSPBOWEYUGHB-UHFFFAOYSA-N
1-(2,4-difluorophenyl)piperazine
2734637
99%
74°C to 76°C
C10H12F2N2
1 g
8620369
1-2,4-difluorophenyl piperazine, 1-2,4-difluorophenyl-piperazine, piperazine, 1-2,4-difluorophenyl, 2,4-difluorophenyl piperazine, pubchem9352, acmc-1ca8h, 2,4-difluorophenylpiperazine, 4-2,4-difluorophenyl piperazine, 1-2,4-difluoro-phenyl-piperazine, piperazine,1-2,4-difluorophenyl
C1CN(CCN1)C2=C(C=C(C=C2)F)F
198.217
198.22
1-(2,4-Difluorophenyl)piperazine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only