accessibility menu, dialog, popup

UserName

FNNH2F

CAS RN 115761-79-0

IUPAC Name: 4-(2,4-difluorophenyl)piperazin-1-ium
Synonyms: 4-(2,4-difluorophenyl)piperazin-1-ium
Molecular Weight (g/mol): 199.22
Molecular Formula: C10H13F2N2
InChi Key: CMCSPBOWEYUGHB-UHFFFAOYSA-O
SMILES: FC1=CC=C(N2CC[NH2+]CC2)C(F)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)