missing translation for 'onlineSavingsMsg'
Learn More

1-(2,4-Difluorophenyl)piperazine, 99%, Thermo Scientific™

Catalog No. AAL1939314
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1939314 25 g
AAL1939303 1 g
AAL1939306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1939314 Supplier Thermo Scientific Chemicals Supplier No. L1939314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 115761-79-0
Molecular Formula C10H12F2N2
Molecular Weight (g/mol) 198.217
MDL Number MFCD00082588
InChI Key CMCSPBOWEYUGHB-UHFFFAOYSA-N
PubChem CID 2734637
IUPAC Name 1-(2,4-difluorophenyl)piperazine
SMILES C1CN(CCN1)C2=C(C=C(C=C2)F)F

Specifications

Melting Point 74°C to 76°C
Boiling Point 108°C to 111°C (0.2 mmHg)
Quantity 25 g
UN Number UN2923
Beilstein 8620369
Sensitivity Air Sensitive
Synonym 1-2,4-difluorophenyl piperazine, 1-2,4-difluorophenyl-piperazine, piperazine, 1-2,4-difluorophenyl, 2,4-difluorophenyl piperazine, pubchem9352, acmc-1ca8h, 2,4-difluorophenylpiperazine, 4-2,4-difluorophenyl piperazine, 1-2,4-difluoro-phenyl-piperazine, piperazine,1-2,4-difluorophenyl
Formula Weight 198.22
Percent Purity 99%
Chemical Name or Material 1-(2,4-Difluorophenyl)piperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only