missing translation for 'onlineSavingsMsg'
Learn More

1-[2-(4-chlorophenyl)-6H-1,3-thiazin-5-yl]ethan-1-one, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB05406EE

Quantity 50g

 View more versions of this product

Catalog No. BTB05406EE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

219539-29-4
251.73
NEKOTKLWPGNIOI-UHFFFAOYSA-N
2799112
CC(=O)C1=CN=C(SC1)C1=CC=C(Cl)C=C1
C12H10ClNOS
MFCD00276113
1-2-4-chlorophenyl-6h-1,3-thiazin-5-yl ethan-1-one, 1-2-4-chlorophenyl-6h-1,3-thiazin-5-yl ethanone, maybridge1_001465, ethanone,1-2-4-chlorophenyl-6h-1,3-thiazin-5-yl
1-[2-(4-chlorophenyl)-6H-1,3-thiazin-5-yl]ethanone

Specifications

1-[2-(4-chlorophenyl)-6H-1, 3-thiazin-5-yl]ethan-1-one
96.5
97%
50g
1-2-4-chlorophenyl-6h-1,3-thiazin-5-yl ethan-1-one, 1-2-4-chlorophenyl-6h-1,3-thiazin-5-yl ethanone, maybridge1_001465, ethanone,1-2-4-chlorophenyl-6h-1,3-thiazin-5-yl
CC(=O)C1=CN=C(SC1)C1=CC=C(Cl)C=C1
251.73
251.74
219539-29-4
100.0
C12H10ClNOS
MFCD00276113
NEKOTKLWPGNIOI-UHFFFAOYSA-N
1-[2-(4-chlorophenyl)-6H-1,3-thiazin-5-yl]ethanone
2799112
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Promotions

Promotions