missing translation for 'onlineSavingsMsg'
Learn More

1-[2-(4-chlorophenyl)-6H-1,3-thiazin-5-yl]ethan-1-one, 97%, Thermo Scientific™

Catalog No. p-3881040
Change view
Click to view available options
Quantity:
1g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB05406DA 1g
BTB05406EA 10g
BTB05406EE 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BTB05406DA Supplier Thermo Scientific Chemicals Supplier No. BTB05406DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 219539-29-4
Molecular Formula C12H10ClNOS
Molecular Weight (g/mol) 251.73
MDL Number MFCD00276113
InChI Key NEKOTKLWPGNIOI-UHFFFAOYSA-N
Synonym 1-2-4-chlorophenyl-6h-1,3-thiazin-5-yl ethan-1-one, 1-2-4-chlorophenyl-6h-1,3-thiazin-5-yl ethanone, maybridge1_001465, ethanone,1-2-4-chlorophenyl-6h-1,3-thiazin-5-yl
PubChem CID 2799112
IUPAC Name 1-[2-(4-chlorophenyl)-6H-1,3-thiazin-5-yl]ethanone
SMILES CC(=O)C1=CN=C(SC1)C1=CC=C(Cl)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1g
Formula Weight 251.74
Percent Purity 97%
Chemical Name or Material 1-[2-(4-chlorophenyl)-6H-1, 3-thiazin-5-yl]ethan-1-one
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.