missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetramethylimidazole 98.0+%, TCI America™

Catalog No. p-7136526
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T097125G 25 g
T0971100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T097125G Supplier TCI America Supplier No. T097125G

Chemical Identifiers

CAS 1739-83-9
Molecular Formula C7H12N2
Molecular Weight (g/mol) 124.187
MDL Number MFCD00059162
InChI Key WLUJHMKCLOIRSK-UHFFFAOYSA-N
PubChem CID 74444
IUPAC Name 1,2,4,5-tetramethylimidazole
SMILES CC1=C(N(C(=N1)C)C)C

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 124.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4,5-Tetramethylimidazole