accessibility menu, dialog, popup

UserName

CH3NCH3NH3CH3C

CAS RN 1739-83-9

IUPAC Name: tetramethyl-1H-imidazole
Synonyms: tetramethylimidazole
Molecular Weight (g/mol): 124.19
Molecular Formula: C7H12N2
InChi Key: WLUJHMKCLOIRSK-UHFFFAOYSA-N
SMILES: CN1C(C)=NC(C)=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results