missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetrakis(tert-butylthio)benzene 98.0+%, TCI America™

Catalog No. T22171G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T22171G Supplier TCI America Supplier No. T22171G
Only null left

Chemical Identifiers

CAS 447463-65-2
Molecular Formula C22H38S4
Molecular Weight (g/mol) 430.786
InChI Key LOCUYYPERSWXJI-UHFFFAOYSA-N
PubChem CID 11812054
IUPAC Name 1,2,4,5-tetrakis(tert-butylsulfanyl)benzene
SMILES CC(C)(C)SC1=CC(=C(C=C1SC(C)(C)C)SC(C)(C)C)SC(C)(C)C

Specifications

Melting Point 156°C
Color White
Quantity 1 g
Formula Weight 430.79
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4,5-Tetrakis(tert-butylthio)benzene