missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetrakis(bromomethyl)benzene, 95%

CAS: 15442-91-8 | C10H10Br4 | 449.79 g/mol

$329.85 - $1332.24

Chemical Identifiers

CAS 15442-91-8
Molecular Formula C10H10Br4
Molecular Weight (g/mol) 449.79
MDL Number MFCD00009664
InChI Key UTXIKCCNBUIWPT-UHFFFAOYSA-N
Synonym 1,2,4,5-tetrakis bromomethyl benzene, 1,2,4,5-tetra bromomethyl benzene, 1,2,4,5-tetrakis-bromomethyl-benzene, benzene, 1,2,4,5-tetrakis bromomethyl, 1,2,4,5-tetrakis bromomethyl-benzene, ksc182m6b, alpha,alpha',alpha,alpha'-tetrabromodurene, 1,2,4,5-tetrakis bromomethyl benzene 5g
PubChem CID 284971
IUPAC Name 1,2,4,5-tetrakis(bromomethyl)benzene
SMILES C1=C(C(=CC(=C1CBr)CBr)CBr)CBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC370920050
SDS View Documents
Thermo Scientific Chemicals
370920050
5 g Glass bottle
Each for $329.85
Only null left
 
AC370920250
SDS View Documents
Thermo Scientific Chemicals
370920250
25 g Glass bottle
Each for $1,332.24
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15442-91-8
449.79
UTXIKCCNBUIWPT-UHFFFAOYSA-N
284971
C1=C(C(=CC(=C1CBr)CBr)CBr)CBr
C10H10Br4
MFCD00009664
1,2,4,5-tetrakis bromomethyl benzene, 1,2,4,5-tetra bromomethyl benzene, 1,2,4,5-tetrakis-bromomethyl-benzene, benzene, 1,2,4,5-tetrakis bromomethyl, 1,2,4,5-tetrakis bromomethyl-benzene, ksc182m6b, alpha,alpha',alpha,alpha'-tetrabromodurene, 1,2,4,5-tetrakis bromomethyl benzene 5g
1,2,4,5-tetrakis(bromomethyl)benzene

Specifications

15442-91-8
100.0
White to Yellow
94% min. (HPLC)
C10H10Br4
MFCD00009664
05, IV, 1080
UTXIKCCNBUIWPT-UHFFFAOYSA-N
1,2,4,5-tetrakis(bromomethyl)benzene
284971
95%
1,2,4,5-Tetrakis(bromomethyl)benzene, 95%
94.0
156°C to 162°C
Authentic
Glass bottle
C6H2(CH2Br)4
5 g
1,2,4,5-tetrakis bromomethyl benzene, 1,2,4,5-tetra bromomethyl benzene, 1,2,4,5-tetrakis-bromomethyl-benzene, benzene, 1,2,4,5-tetrakis bromomethyl, 1,2,4,5-tetrakis bromomethyl-benzene, ksc182m6b, alpha,alpha',alpha,alpha'-tetrabromodurene, 1,2,4,5-tetrakis bromomethyl benzene 5g
C1=C(C(=CC(=C1CBr)CBr)CBr)CBr
449.79
449.79
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only