missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetrafluorobenzene 99.0+%, TCI America™

Catalog No. p-7136562
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10295G 5 g
T102925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10295G Supplier TCI America Supplier No. T10295G
Only null left

Chemical Identifiers

CAS 327-54-8
Molecular Formula C6H2F4
Molecular Weight (g/mol) 150.076
MDL Number MFCD00000307
InChI Key SDXUIOOHCIQXRP-UHFFFAOYSA-N
Synonym benzene, 1,2,4,5-tetrafluoro, 2,3,5,6-tetrafluorobenzene, pubchem1063, acmc-209hvc, 1,4,5-tetrafluorobenzene, 2,5,6-tetrafluorobenzene, 2,3,5,6-tetrafluorophenyl, ksc496i2b, benzene,2,4,5-tetrafluoro, 1,2,4,5-tetrafluoro-benzene
PubChem CID 9474
IUPAC Name 1,2,4,5-tetrafluorobenzene
SMILES C1=C(C(=CC(=C1F)F)F)F

Specifications

Color Colorless
Boiling Point 90°C
Quantity 5 g
UN Number 1993
Formula Weight 150.08
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,4,5-Tetrafluorobenzene