missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetrafluorobenzene, 99%

CAS: 327-54-8 | C6H2F4 | 150.076 g/mol

Supplier:  Thermo Scientific Chemicals A1781514

 View more versions of this product

Catalog No. AAA1781514


Only null left
Explore available promotions

Description

Description

It is employed as intermediate in organic synthesis. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate in organic synthesis. It is also used as pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources.
Specifications

Specifications

1,2,4,5-Tetrafluorobenzene
3°C to 5°C
90°C
C6H2F4
MFCD00000307
UN2387
benzene, 1,2,4,5-tetrafluoro, 2,3,5,6-tetrafluorobenzene, pubchem1063, acmc-209hvc, 1,4,5-tetrafluorobenzene, 2,5,6-tetrafluorobenzene, 2,3,5,6-tetrafluorophenyl, ksc496i2b, benzene,2,4,5-tetrafluoro, 1,2,4,5-tetrafluoro-benzene
SDXUIOOHCIQXRP-UHFFFAOYSA-N
1,2,4,5-tetrafluorobenzene
9474
99%
327-54-8
1.43
16°C (60°F)
1.407
25 g
1909052
Not miscible or difficult to mix in water.
C1=C(C(=CC(=C1F)F)F)F
150.076
150.07

Chemical Identifiers

327-54-8
150.076
SDXUIOOHCIQXRP-UHFFFAOYSA-N
9474
C1=C(C(=CC(=C1F)F)F)F
C6H2F4
MFCD00000307
benzene, 1,2,4,5-tetrafluoro, 2,3,5,6-tetrafluorobenzene, pubchem1063, acmc-209hvc, 1,4,5-tetrafluorobenzene, 2,5,6-tetrafluorobenzene, 2,3,5,6-tetrafluorophenyl, ksc496i2b, benzene,2,4,5-tetrafluoro, 1,2,4,5-tetrafluoro-benzene
1,2,4,5-tetrafluorobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROBENZENE

EINECSNumber : 206-319-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only