missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetrafluorobenzene, 99%

Catalog No. AAA1781506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1781506 5 g
AAA1781514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1781506 Supplier Thermo Scientific Chemicals Supplier No. A1781506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 327-54-8 | C6H2F4 | 150.076 g/mol

It is employed as intermediate in organic synthesis. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate in organic synthesis. It is also used as pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources.

Chemical Identifiers

CAS 327-54-8
Molecular Formula C6H2F4
Molecular Weight (g/mol) 150.076
MDL Number MFCD00000307
InChI Key SDXUIOOHCIQXRP-UHFFFAOYSA-N
Synonym benzene, 1,2,4,5-tetrafluoro, 2,3,5,6-tetrafluorobenzene, pubchem1063, acmc-209hvc, 1,4,5-tetrafluorobenzene, 2,5,6-tetrafluorobenzene, 2,3,5,6-tetrafluorophenyl, ksc496i2b, benzene,2,4,5-tetrafluoro, 1,2,4,5-tetrafluoro-benzene
PubChem CID 9474
IUPAC Name 1,2,4,5-tetrafluorobenzene
SMILES C1=C(C(=CC(=C1F)F)F)F

Specifications

Melting Point 3°C to 5°C
Density 1.43
Boiling Point 90°C
Flash Point 16°C (60°F)
Refractive Index 1.407
Quantity 5 g
UN Number UN2387
Beilstein 1909052
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 150.07
Percent Purity 99%
Chemical Name or Material 1,2,4,5-Tetrafluorobenzene
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROBENZENE
EINECSNumber 206-319-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only