missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,4,5-Tetracyanobenzene 98.0+%, TCI America™
$139.81 - $561.08
Chemical Identifiers
| CAS | 712-74-3 |
|---|---|
| Molecular Formula | C10H2N4 |
| Molecular Weight (g/mol) | 178.154 |
| MDL Number | MFCD00043481 |
| InChI Key | FAAXSAZENACQBT-UHFFFAOYSA-N |
| Synonym | 1,2,4,5-tetracyanobenzene, 1,2,4,5-benzenetetracarbonitrile, tetracyanobenzene, pyromellitic nitrile, pyromellitotetranitrile, pyromellitic tetranitrile, pyromellitic acid tetranitrile, 1,2,4,5-benzenetetranitrile, pyromellitonitrile, s-tetracyanobenzene |
| PubChem CID | 12838 |
| IUPAC Name | benzene-1,2,4,5-tetracarbonitrile |
| SMILES | C1=C(C(=CC(=C1C#N)C#N)C#N)C#N |
Chemical Identifiers
| 712-74-3 | |
| 178.154 | |
| FAAXSAZENACQBT-UHFFFAOYSA-N | |
| 12838 | |
| C1=C(C(=CC(=C1C#N)C#N)C#N)C#N |
| C10H2N4 | |
| MFCD00043481 | |
| 1,2,4,5-tetracyanobenzene, 1,2,4,5-benzenetetracarbonitrile, tetracyanobenzene, pyromellitic nitrile, pyromellitotetranitrile, pyromellitic tetranitrile, pyromellitic acid tetranitrile, 1,2,4,5-benzenetetranitrile, pyromellitonitrile, s-tetracyanobenzene | |
| benzene-1,2,4,5-tetracarbonitrile |
Specifications
| 712-74-3 | |
| White-Yellow | |
| MFCD00043481 | |
| 3439 | |
| FAAXSAZENACQBT-UHFFFAOYSA-N | |
| benzene-1,2,4,5-tetracarbonitrile | |
| 12838 | |
| ≥98.0% (HPLC,N) | |
| 1,2,4,5-Tetracyanobenzene |
| 265°C | |
| C10H2N4 | |
| 1 g | |
| 1,2,4,5-tetracyanobenzene, 1,2,4,5-benzenetetracarbonitrile, tetracyanobenzene, pyromellitic nitrile, pyromellitotetranitrile, pyromellitic tetranitrile, pyromellitic acid tetranitrile, 1,2,4,5-benzenetetranitrile, pyromellitonitrile, s-tetracyanobenzene | |
| C1=C(C(=CC(=C1C#N)C#N)C#N)C#N | |
| 178.154 | |
| 178.15 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : DB9100000
TSCA : No