accessibility menu, dialog, popup

UserName

CAS RN 712-74-3

NNNN

CAS RN 712-74-3

IUPAC Name: benzene-1,2,4,5-tetracarbonitrile
Synonyms: 1,2,4,5-tetracyanobenzene
Molecular Weight (g/mol): 178.15
Molecular Formula: C10H2N4
InChi Key: FAAXSAZENACQBT-UHFFFAOYSA-N
SMILES: N#CC1=CC(C#N)=C(C=C1C#N)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results