missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,4,5-Tetrabromobenzene 97.0+%, TCI America™
Supplier: TCI America T003125G
Specifications
| 1,2,4,5-Tetrabromobenzene | |
| 180°C | |
| MFCD00000063 | |
| benzene, 1,2,4,5-tetrabromo, 2,3,5,6-tetrabromobenzene, unii-m25dk66lcz, m25dk66lcz, pubchem4448, 1,4,5-tetrabromobenzene, 2,5,6-tetrabromobenzene, acmc-1b6z1, benzene,2,4,5-tetrabromo, benzene,1,2,4,5-tetrabromo | |
| BrC1=CC(Br)=C(Br)C=C1Br | |
| 393.70 | |
| 393.70 | |
| Crystalline Powder |
| 636-28-2 | |
| C6H2Br4 | |
| 25 g | |
| QCKHVNQHBOGZER-UHFFFAOYSA-N | |
| 1,2,4,5-tetrabromobenzene | |
| 12486 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 636-28-2 | |
| 393.70 | |
| QCKHVNQHBOGZER-UHFFFAOYSA-N | |
| 1,2,4,5-tetrabromobenzene |
| C6H2Br4 | |
| MFCD00000063 | |
| 12486 | |
| BrC1=CC(Br)=C(Br)C=C1Br |
Safety and Handling
TSCA : Yes