Learn More
1,2,4,5-Tetrabromobenzene, 97%
CAS: 636-28-2 | C6H2Br4 | 393.70 g/mol
$469.46 - $469.46
Chemical Identifiers
| CAS | 636-28-2 |
|---|---|
| Molecular Formula | C6H2Br4 |
| Molecular Weight (g/mol) | 393.70 |
| MDL Number | MFCD00000063 |
| InChI Key | QCKHVNQHBOGZER-UHFFFAOYSA-N |
| PubChem CID | 12486 |
| IUPAC Name | 1,2,4,5-tetrabromobenzene |
| SMILES | BrC1=CC(Br)=C(Br)C=C1Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC352401000
|
Thermo Scientific Chemicals
352401000 |
100 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 636-28-2 | |
| 393.70 | |
| QCKHVNQHBOGZER-UHFFFAOYSA-N | |
| 1,2,4,5-tetrabromobenzene |
| C6H2Br4 | |
| MFCD00000063 | |
| 12486 | |
| BrC1=CC(Br)=C(Br)C=C1Br |
Specifications
| 636-28-2 | |
| 100.0 | |
| Beige to Brown | |
| 96% min. (GC) | |
| C6H2Br4 | |
| MFCD00000063 | |
| benzene, 1,2,4,5-tetrabromo, 2,3,5,6-tetrabromobenzene, unii-m25dk66lcz, m25dk66lcz, pubchem4448, 1,4,5-tetrabromobenzene, 2,5,6-tetrabromobenzene, acmc-1b6z1, benzene,2,4,5-tetrabromo, benzene,1,2,4,5-tetrabromo | |
| QCKHVNQHBOGZER-UHFFFAOYSA-N | |
| 1,2,4,5-tetrabromobenzene | |
| 12486 | |
| 97% | |
| 1,2,4,5-Tetrabromobenzene, 97% |
| 96.0 | |
| 177.0°C to 182.0°C | |
| Authentic | |
| Glass bottle | |
| C6H2Br4 | |
| 100 g | |
| Solubility in water: insoluble. | |
| BrC1=CC(Br)=C(Br)C=C1Br | |
| 393.70 | |
| 393.7 | |
| Crystalline Powder, Crystals or Needles |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause long lasting harmful effects to aquatic life.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 211-253-3
TSCA : TSCA
RUO – Research Use Only