missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetrabromobenzene, 94%

Catalog No. AAL1353909
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1353909 10 g
AAL1353918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1353909 Supplier Thermo Scientific Chemicals Supplier No. L1353909

CAS: 636-28-2 | C6H2Br4 | 393.70 g/mol

1,2,4,5-Tetrabromobenzene is used in the synthesis of diethynyltriptycene-linked dipyridyl ligands, in the generation of dibenzyne. It is also used to produce 6,7-dibromo-1,4-epoxy-1,4-dihydronaphthalene at temperature of -23°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4,5-Tetrabromobenzene is used in the synthesis of diethynyltriptycene-linked dipyridyl ligands, in the generation of dibenzyne. It is also used to produce 6,7-dibromo-1,4-epoxy-1,4-dihydronaphthalene at temperature of -23°C.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 636-28-2
Molecular Formula C6H2Br4
Molecular Weight (g/mol) 393.70
MDL Number MFCD00000063
InChI Key QCKHVNQHBOGZER-UHFFFAOYSA-N
Synonym benzene, 1,2,4,5-tetrabromo, 2,3,5,6-tetrabromobenzene, unii-m25dk66lcz, m25dk66lcz, pubchem4448, 1,4,5-tetrabromobenzene, 2,5,6-tetrabromobenzene, acmc-1b6z1, benzene,2,4,5-tetrabromo, benzene,1,2,4,5-tetrabromo
PubChem CID 12486
IUPAC Name 1,2,4,5-tetrabromobenzene
SMILES BrC1=CC(Br)=C(Br)C=C1Br

Specifications

Melting Point 175°C to 180°C
Quantity 10 g
Beilstein 1365830
Solubility Information Insoluble in water.
Formula Weight 393.7
Percent Purity 94%
Chemical Name or Material 1,2,4,5-Tetrabromobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-253-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only