missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Tris(2-cyanoethoxy)propane 97.0+%, TCI America™

Catalog No. T052525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T052525G 25 g
T0525500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T052525G Supplier TCI America Supplier No. T052525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2465-93-2
Molecular Formula C12H17N3O3
Molecular Weight (g/mol) 251.286
MDL Number MFCD00019868
InChI Key ALGVJKNIAOBBBJ-UHFFFAOYSA-N
Synonym 1,2,3-tris 2-cyanoethoxy propane, propanenitrile, 3,3',3-1,2,3-propanetriyltris oxy tris, 3,3',3-propane-1,2,3-triyltrioxytripropiononitrile, tris .beta.-cyanoethoxy propane, glycerol tris 2-cyanoethyl ether, propionitrile, 3,3',3-1,2,3-propanetriyltrioxy tri, 3-2,3-bis 2-cyanoethoxy propoxy propanenitrile, 3-1,3-bis 2-cyanoethoxy propan-2-yl oxy propanenitrile, propanenitrile,3,3',3-1,2,3-propanetriyltris oxy tris
PubChem CID 75572
IUPAC Name 3-[2,3-bis(2-cyanoethoxy)propoxy]propanenitrile
SMILES C(COCC(COCCC#N)OCCC#N)C#N

Specifications

Color Yellow
Quantity 25 g
UN Number 3276
Formula Weight 251.29
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,3-Tris(2-cyanoethoxy)propane