accessibility menu, dialog, popup

UserName

NOONON

CAS RN 2465-93-2

IUPAC Name: 3-{[1,3-bis(2-cyanoethoxy)propan-2-yl]oxy}propanenitrile
Synonyms: 3-{[1,3-bis(2-cyanoethoxy)propan-2-yl]oxy}propanenitrile
Molecular Weight (g/mol): 251.29
Molecular Formula: C12H17N3O3
InChi Key: ALGVJKNIAOBBBJ-UHFFFAOYSA-N
SMILES: N#CCCOCC(COCCC#N)OCCC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results