missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trimethyl-3H-benzo[e]indole 98.0+%, TCI America™

Catalog No. p-7137587
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31195G 5 g
T311925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31195G Supplier TCI America Supplier No. T31195G

Chemical Identifiers

CAS 881219-73-4
Molecular Formula C15H15N
Molecular Weight (g/mol) 209.29
MDL Number MFCD06651515
InChI Key KQMROBBTYFXFOV-UHFFFAOYSA-N
Synonym 1,2,3-Trimethylnaphtho[1,2-d]pyrrole
PubChem CID 16439627
IUPAC Name 1,2,3-trimethyl-3H-benzo[e]indole
SMILES CN1C(C)=C(C)C2=C1C=CC1=CC=CC=C21

Specifications

Quantity 5 g
Formula Weight 209.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3-Trimethyl-3H-benzo[e]indole