accessibility menu, dialog, popup

UserName

CAS RN 881219-73-4

H3CNCH3CH3

CAS RN 881219-73-4

IUPAC Name: 1,2,3-trimethyl-3H-benzo[e]indole
Synonyms: 1,2,3-trimethylbenzo[e]indole
Molecular Weight (g/mol): 209.29
Molecular Formula: C15H15N
InChi Key: KQMROBBTYFXFOV-UHFFFAOYSA-N
SMILES: CN1C(C)=C(C)C2=C1C=CC1=CC=CC=C21
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results