missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trimethoxybenzene 99.0+%, TCI America™

Catalog No. p-7136608
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T110225G 25 g
T1102500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T110225G Supplier TCI America Supplier No. T110225G
Only null left

Chemical Identifiers

CAS 634-36-6
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00008358
InChI Key CRUILBNAQILVHZ-UHFFFAOYSA-N
Synonym methylsyringol, pyrogallol trimethyl ether, trimethoxybenzene, benzene, 1,2,3-trimethoxy, tri-o-methylpyrogallol, benzene, trimethoxy, unii-mre1o894fg, 1,2,3-trimethoxy benzene, mre1o894fg, 3,4,5-trimethoxybenzene
PubChem CID 12462
ChEBI CHEBI:86529
IUPAC Name 1,2,3-trimethoxybenzene
SMILES COC1=C(C(=CC=C1)OC)OC

Specifications

Melting Point 45°C
Color White
Boiling Point 235°C
Quantity 25 g
Formula Weight 168.19
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3-Trimethoxybenzene