missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trimethoxybenzene, 98%

Catalog No. p-206233
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160210250 25 g Glass bottle
AC160211000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC160210250 Supplier Thermo Scientific Chemicals Supplier No. 160210250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 634-36-6 | C9H12O3 | 168.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 634-36-6
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
MDL Number MFCD00008358
InChI Key CRUILBNAQILVHZ-UHFFFAOYSA-N
Synonym methylsyringol, pyrogallol trimethyl ether, trimethoxybenzene, benzene, 1,2,3-trimethoxy, tri-o-methylpyrogallol, benzene, trimethoxy, unii-mre1o894fg, 1,2,3-trimethoxy benzene, mre1o894fg, 3,4,5-trimethoxybenzene
PubChem CID 12462
ChEBI CHEBI:86529
IUPAC Name 1,2,3-trimethoxybenzene
SMILES COC1=C(C(=CC=C1)OC)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 42.0°C to 45.0°C
Color Beige to White
Density 1.1120g/mL
Boiling Point 241.0°C
Flash Point 112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H3(OCH3)3
Quantity 25 g
Beilstein 06, 1081
Specific Gravity 1.112
Solubility Information Solubility in water: insoluble
Formula Weight 168.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1, 2, 3-Trimethoxybenzene, 98%
Show More Show Less
EINECSNumber 211-207-2
TSCA TSCA

RUO – Research Use Only