missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trimethoxybenzene, 98+%

Catalog No. p-7022000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1457214 25 g
AAA1457222 100 g
AAA1457236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1457214 Supplier Thermo Scientific Chemicals Supplier No. A1457214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 634-36-6 | C9H12O3 | 168.192 g/mol

1,2,3-Trimethoxybenzene was used to study the effect of solvent on photo induced electron-transfer reactions, on condensation with 2,4-diamino-5-(hydroxymethyl)pyrimidine yields 2,4-diamino-5-(2,3,4-trimethoxybenzyl)pyrimidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3-Trimethoxybenzene was used to study the effect of solvent on photo induced electron-transfer reactions, on condensation with 2,4-diamino-5-(hydroxymethyl)pyrimidine yields 2,4-diamino-5-(2,3,4-trimethoxybenzyl)pyrimidine.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 634-36-6
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00008358
InChI Key CRUILBNAQILVHZ-UHFFFAOYSA-N
Synonym methylsyringol, pyrogallol trimethyl ether, trimethoxybenzene, benzene, 1,2,3-trimethoxy, tri-o-methylpyrogallol, benzene, trimethoxy, unii-mre1o894fg, 1,2,3-trimethoxy benzene, mre1o894fg, 3,4,5-trimethoxybenzene
PubChem CID 12462
ChEBI CHEBI:86529
IUPAC Name 1,2,3-trimethoxybenzene
SMILES COC1=C(C(=CC=C1)OC)OC

Specifications

Melting Point 43°C to 45°C
Density 1.112
Boiling Point 241°C
Flash Point 112°C (233°F)
Quantity 25 g
Beilstein 1910422
Solubility Information Insoluble in water.
Formula Weight 168.19
Percent Purity ≥98%
Chemical Name or Material 1,2,3-Trimethoxybenzene
EINECSNumber 211-207-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only