missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3-Trimethoxybenzene, 98+%
Description
1,2,3-Trimethoxybenzene was used to study the effect of solvent on photo induced electron-transfer reactions, on condensation with 2,4-diamino-5-(hydroxymethyl)pyrimidine yields 2,4-diamino-5-(2,3,4-trimethoxybenzyl)pyrimidine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2,3-Trimethoxybenzene was used to study the effect of solvent on photo induced electron-transfer reactions, on condensation with 2,4-diamino-5-(hydroxymethyl)pyrimidine yields 2,4-diamino-5-(2,3,4-trimethoxybenzyl)pyrimidine.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.
1,2,3-Trimethoxybenzene was used to study the effect of solvent on photo induced electron-transfer reactions, on condensation with 2,4-diamino-5-(hydroxymethyl)pyrimidine yields 2,4-diamino-5-(2,3,4-trimethoxybenzyl)pyrimidine.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 43°C to 45°C |
| Density | 1.112 |
| Boiling Point | 241°C |
| Flash Point | 112°C (233°F) |
| Quantity | 100 g |
| Beilstein | 1910422 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 168.19 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 1,2,3-Trimethoxybenzene |
Safety and Handling
| EINECSNumber | 211-207-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only