missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3-Thiadiazole-4-carbaldehyde, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 1g |
| Formula Weight | 114.13 |
| Percent Purity | 97% |
| Chemical Name or Material | 1, 2, 3-Thiadiazole-4-carbaldehyde |